Fluorescent western blot stripping buffer, Restore™
Restore™ Fluorescent Western Blot Stripping Buffer, 20 ml, 1
Catalog # PIER62299
Supplier: Thermo Fisher Scientific
Fluorescent western blot stripping buffer, Restore™
Catalog # PIER62299
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Restore™ Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from western blots.
- Strip blots in only 15 minutes at room temperature
- Reprobe the same PVDF membrane for multiple targets
- Effectively strips blots the first time
- No need to run new gels and prepare a new blot
Specifications
Specifications
- Pk:1
- Description:Restore™ Fluorescent Western Blot Stripping Buffer, 20 ml