Orders LinkContactUsLinkComponent
Gram Negative lysis buffer
Gram Negative lysis buffer
Gram negative lysis buffer, 250 mL
Catalog # 786-567
Supplier:   G-Biosciences
Gram Negative lysis buffer
Catalog # 786-567
Supplier:  G-Biosciences
CAS Number:  
Gram Negative lysis buffer
Gram Negative lysis buffer
Gram negative lysis buffer, 250 mL
CAS Number:  
Price
101,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Gram-negative lysis buffer is an extraction buffer for soluble proteins from Gram-negative bacteria. It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins. This buffer has been tested for use with several widely used bacteria including E.coli strains.
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    Gram negative lysis buffer