Order Entry
Germany
Orders LinkContactUsLinkComponent
Hybridisation buffers

Hybridisation buffers

Denhardt solution [100X], 50 mL
Catalog # 786-525
Supplier:   G-Biosciences
Hybridisation buffers
Catalog # 786-525
Supplier:  G-Biosciences
CAS Number:  
Hybridisation buffers
Hybridisation buffers

Denhardt solution [100X], 50 mL

CAS Number:  
Price
149,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

The hybridisation buffers like Church & Gilbert’s, Denhardt solution, hybridisation denaturing solution, hybridisation neutralising solution, SSC, SSPE, TMAC Hyb solution and TNT Buffer are used in wide range of applications. Church & Gilbert’s Hybridization Buffer is prepared from molecular biology grade SDS (sodium dodecyl sulfate), monosodium phosphate (NaHPO₄), EDTA (ethylenediaminetetracetic acid) and bovine serum albumin (BSA) Fraction V using DNase, RNase and Protease-free water. It consists of 1mM EDTA, 1% BSA, 0,5M sodium phosphate monobasic, and 7% SDS (at a pH of 7,2).

Denhardt solution is suitable for nucleic acid hybridisation. Denhardt's solution is a mi×ture of high-molecular weight polymers capable of saturating non-specific binding sites and artificially increasing the concentration of available probe. It consists of 2% Ficoll 400, 300mM NaCl, 2% polyvinylpyrrolidone, and 2% BSA.
The SSPE or or Saline-Sodium phosphate-EDTA buffer is a general hybridisation buffer that consists of 0,2 M phosphate buffer (at a pH of 7,4), 2,98 M NaCl, and 0,02 M EDTA.
The TMAC hybridisation Solution consists of 3 M Tetramethyl ammonium chloride, 50mm Tris-HCl (at a pH of 8,0), 1 mm EDTA (at a pH of 8,0), and 0,1% SDS.
The TNT Buffer is a general molecular biology and hybridisation buffer that consists of 0,1 M Tris-HCl (at a pH of 7,5), 0,15 M NaCl, and 0,05% Tween®-20.

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Denhardt solution [100X]

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.