Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution for chlorine determination

Buffer solution for chlorine determination

4,0, Acetate, 100 ml
Catalog # HACH1490932
Supplier:   Hach
Buffer solution for chlorine determination
Catalog # HACH1490932
Supplier:  Hach
CAS Number:  
Buffer solution for chlorine determination
Buffer solution for chlorine determination

4,0, Acetate, 100 ml

CAS Number:  
Price
34,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.

Specifications

Specifications

  • pH buffer type:
    Acetate
  • Pk:
    100 ml
  • pH value:
    4,0

Specifications