Order Entry
Germany
Orders LinkContactUsLinkComponent
Safety Bottle Container

Safety Bottle Container

Silicone seal for transport container, Red, Container with Ø 130 mm
Catalog # BOCH18300SS
Supplier:   BOCHEM
Safety Bottle Container
Catalog # BOCH18300SS
Supplier:  BOCHEM
Safety Bottle Container
Safety Bottle Container

Silicone seal for transport container, Red, Container with Ø 130 mm

Price
22,40 €
Log in for availability
1 * 5 ST
Product Details & Documents

Safety transport container, made of stainless steel, for laboratory bottles.

  • For bottles 50 to 1000 ml
  • With hinge, lock and silicone seal
  • Good chemical resistance
Specifications
  • For:
    Container with Ø 130 mm
  • Colour:
    Red
  • Description:
    Silicone seal for transport container
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.