Orders LinkContactUsLinkComponent
Screw Caps
Screw Caps
GL 25
Artikelnummer HECH41326025
Screw Caps
Artikelnummer HECH41326025
Screw Caps
GL 25
Preis
3,20 €
Log in for availability
1 * 10 ST
Product Details & Documents
PP, black.
  • Very good chemical resistance
  • Heat-resistant up to 110 °C
  • Only suitable for bottles 215-2814 to 215-2819
Specifications
  • Thread:
    GL 25
  • Seq:
    2