ContactUsLinkComponent
Tubing, Dual Layer PEEK, 1/16'' x 0.75 mm ID, solid green outside, natural inside, 25 m/pkg 1 * 25 m
Catalog # JOURJR-TD-6011-M25
Supplier:  VICI JOUR RESEARCH
Tubing, Dual Layer PEEK, 1/16'' x 0.75 mm ID, solid green outside, natural inside, 25 m/pkg 1 * 25 m
Catalog # JOURJR-TD-6011-M25
Supplier:  VICI JOUR RESEARCH
Frequently Bought Together