Orders LinkContactUsLinkComponent
Caps
Caps
Cap kit, includes one cap, with fittings, one 1 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter, GL 45, 1 port for 1/16" tubing, 3 ports for 1/8" and 1/16" OD tubing, PP
Catalog # JOURJR-S-11103
Supplier:   VICI JOUR RESEARCH
Caps
Catalog # JOURJR-S-11103
Supplier:  VICI JOUR RESEARCH
Caps
Cap kit, includes one cap, with fittings, one 1 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter, GL 45, 1 port for 1/16" tubing, 3 ports for 1/8" and 1/16" OD tubing, PP
Price
341,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifications
  • Thread:
    GL 45
  • Description:
    Cap kit, includes one cap, with fittings, one 1 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter
  • Material:
    PP
  • Seq:
    15
  • Connector:
    1 port for 1/16" tubing, 3 ports for 1/8" and 1/16" OD tubing