Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
250 ml, 29/32 NS
Catalog # 531-0833
Supplier:   Lenz Laborglas
Rotary evaporating flasks
Catalog # 531-0833
Supplier:  Lenz Laborglas
Rotary evaporating flasks
Rotary evaporating flasks
250 ml, 29/32 NS
Price
97,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Documents
Certificates
Search by Lot Number
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml
  • Seq:
    15