Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® Basic, Bouffant Caps, Latex-Free

VWR® Basic, Bouffant Caps, Latex-Free

White, 530 mm
Catalog # 113-1459
Supplier:   VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
Catalog # 113-1459
Supplier:  VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free

White, 530 mm

Price
70,40 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Available in a wide range of colours

Weight: 10 g/m²

Packaging: 100 pieces per bag, 10 bags per case.

Specifications
  • Ø:
    530 mm
  • Colour:
    White
  • Seq:
    15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.