Orders LinkContactUsLinkComponent
Caps
Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, GL 45, 1 port for 1/16" tubing, 1 port for 1/8" and 1/16" OD tubing, PP
Catalog # JOURJR-S-11001
Supplier:   VICI JOUR RESEARCH
Caps
Catalog # JOURJR-S-11001
Supplier:  VICI JOUR RESEARCH
Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, GL 45, 1 port for 1/16" tubing, 1 port for 1/8" and 1/16" OD tubing, PP
Price
91,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifications
  • Thread:
    GL 45
  • Description:
    Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter
  • Material:
    PP
  • Seq:
    01
  • Connector:
    1 port for 1/16" tubing, 1 port for 1/8" and 1/16" OD tubing