Orders LinkContactUsLinkComponent
Screw Caps
Screw Caps
GL 18
Catalog # HECH41326018
Screw Caps
Catalog # HECH41326018
Screw Caps
GL 18
Price
1,90 €
Log in for availability
1 * 10 ST
Product Details & Documents
PP, black.
  • Very good chemical resistance
  • Heat-resistant up to 110 °C
  • Only suitable for bottles 215-2814 to 215-2819
Specifications
  • Thread:
    GL 18
  • Seq:
    1