Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
V-bottom, clear, non sterile, 40 - 200 µl, -, 234 µl
Catalog # 738-0208
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 96-well
Catalog # 738-0208
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 96-well
Microplates, 96-well
V-bottom, clear, non sterile, 40 - 200 µl, -, 234 µl
Price
306,00 €
Log in for availability
1 * 400 ST
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications
  • Recommended working volume:
    40 - 200 µl
  • Sterile:
    -
  • Description:
    V-bottom, clear, non sterile
  • Seq:
    08
  • Total volume:
    234 µl
  • Cat. No.:
    738-0208
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact