FORMALDEHYDE SOLUTION 1 * 100 mL
Catalog # REGE1039101
Supplier: REAGECON
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethyl Acetate, 99.5% for synthesis 1 * 5 L
Supplier: PanReac AppliChem
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
KUPFERSULFAT 10 % WÄSSRIG 1 * 500 mL
Supplier: MARKET SOURCE PART PROCESS
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Cyclopentyl methyl ether (CPME) ≥99.9%
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific