Storage Bins
Red, 165×105×75 mm, PP
Catalog # SEMA2791
Supplier: SEMADENI
Jump to:
Product Details & Documents
Made of PP.
- Secure stacking
- Made of high quality material and are dimensionally stable
- Supporting edge and smooth inner surfaces for easy cleaning
Storage bins with large labelling field for content description.
Specifications
- Colour:Red
- Material:PP
- Seq:01
- W×D×H:165×105×75 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



