Methylcyclohexane
155 kg
Catalog # OVER8737941-155
Marketsource
Supplier: OVERLACK CHEMISCHE PRODUKTE
Methylcyclohexane
Catalog # OVER8737941-155
Supplier: OVERLACK CHEMISCHE PRODUKTE
Marketsource
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126 °C
- Density:0.7693 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:203-624-3
- UN:2296
- ADR:3,II
Specifications
Specifications
- Pk:155 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



