Nebulisers, PFA-ST/ST3 MicroFlow™
PFA-ST self-aspirating nebuliser
Catalog # PERCN8122192
Supplier: PerkinElmer
Jump to:
Product Details & Documents
The PFA-ST MicroFlow nebuliser is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe.
- Chemically resistant ideal for strong acids, alkalis and organics
- Direct analysis of volatile and non-volatile organic solvents
- Longer lifetime than glass or quartz nebulisers
- With external 1/4 - 28 threaded connector
Exchangeable uptake capillaries allow one nebuliser to be used at different self-aspiration rates from 20 to 700 μl/min. Can be pumped from <0,1 to 3,0 ml/min.
PFA-ST3 designed for superior clog resistance and higher sensitivity compared to the standard PFA nebuliser.
PFA MicroFlow ideal for strong acids, high solids, alkalis, organic solvents, soils and high salt solutions.
Accessories information: Internal standard addition tee and FAST valve nebulizer lines are available.
Specifications
- Description:PFA-ST self-aspirating nebuliser
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- FAST valve line for PFA-ST nebuliser (0,25 mm I.Ø), green, attaches to port #3FAST valve line for PFA-ST nebuliser (0,25 mm I.Ø), green, attaches to port #3PERCN0777287Log in for availability1 * 1 ST - 225,00 €
Product Information
- Catalog #AvailabilityPrice
- Internal standard addition tee for ESI ST nebuliser (0,25 mm I.Ø), green, attaches to port #3Internal standard addition tee for ESI ST nebuliser (0,25 mm I.Ø), green, attaches to port #3PERCN0777294Log in for availability1 * 1 ST - 960,00 €
Product Information
- Catalog #AvailabilityPrice
- Internal standard addition tee for ESI ST nebuliser (0,5 mm I.Ø), orange, attaches to port #3Internal standard addition tee for ESI ST nebuliser (0,5 mm I.Ø), orange, attaches to port #3PERCN0777295Log in for availability1 * 1 ST - 948,00 €