Order Entry
Germany
Orders LinkContactUsLinkComponent
Accessories

Accessories

Micro glass tube, 50 mm, 0,55 ml
Catalog # KRUEPRG-50-M
Supplier:   A.KRÜSS Optronic
Accessories
Catalog # KRUEPRG-50-M
Supplier:  A.KRÜSS Optronic
Accessories

Micro glass tube, 50 mm, 0,55 ml

Price
1 010,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Micro glass tube, 50 mm, 0,55 ml