Pipette tips
VistaBulk™, 5 ml, -, Bulk, 250 tips/bag
Catalog # VIST4058-5000
Supplier: VISTALAB TECHNOLOGIES
Jump to:
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
- Packed:Bulk, 250 tips/bag
- Sterile:-
- Volume:5 ml
- Tip style:VistaBulk™