Orders LinkContactUsLinkComponent
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
medium length, B, 45/40 NS, 29/32 NS
Catalog # COWI021.2945
Supplier:   COWIE
Adapters, PTFE, socket on cone
Catalog # COWI021.2945
Supplier:  COWIE
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
medium length, B, 45/40 NS, 29/32 NS
Price
36,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of pure PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Fully adjustable with replacement Viton® seal
  • Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Specifications
  • Cone size:
    45/40 NS
  • Socket size:
    29/32 NS
  • Type:
    medium length, B
  • Seq:
    12