Orders LinkContactUsLinkComponent
Nutrient medium, Biosart® 100
Nutrient medium, Biosart® 100
Wallerstein differential, Bacteria in fermentation processes, 50 Ampoul
Catalog # 710-0856
Supplier:   Sartorius
Nutrient medium, Biosart® 100
Catalog # 710-0856
Supplier:  Sartorius
CAS Number:  
Nutrient medium, Biosart® 100
Nutrient medium, Biosart® 100
Wallerstein differential, Bacteria in fermentation processes, 50 Ampoul
CAS Number:  
Price
125,00 €
Log in for availability
1 * 50 Ampull
Product Details & Documents
These ready to use ampoules are pre-sterilised and contain a membrane filter and pad. They have been specially designed for performing microbiological analyses on pharmaceuticals, cosmetics, food and drink, water and other liquids. To be used with Biosart® 100 Monitors. When stored in appropriate conditions (4 °C), the medium has a shelf life of up to 12 months (except for Endo). The formula ensures optimum colony growth on membranes and complies with international standards, including International Pharmacopoeias, DIN and ISO standards, American Standards for Water and Foods, mineral water regulations and recommendations for the food and drink industry.
  • Pre-sterilised nutrient medium with quality certificate for each batch
  • Consistently high recovery rates
  • Ready for use and easy to handle, avoids use of glass ampoules
  • Long shelf life
Delivery information: Pre-sterilised Biosart® 100 nutrient medium in 2.5 ml plastic ampoules. Each pack contains 50 plastic ampoules and a batch quality certificate.
Documents
Specifications
Specifications
  • Pk:
    50 Ampoul
  • Description:
    Wallerstein differential
  • For:
    Bacteria in fermentation processes