Orders LinkContactUsLinkComponent
Screw caps
Screw caps
S 28
Catalog # VITL102697
Supplier:   VITLAB
Screw caps
Catalog # VITL102697
Supplier:  VITLAB
Screw caps
Screw caps
S 28
Price
28,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
PFA, transparent
  • Especially suitable for use in trace analysis
  • Ideal for sensitive and valuable samples
  • High thermal stability, from –200 °C to +260 °C, autoclavable
  • Chemical inertness against nearly all chemicals
  • Easy to clean
For sealing all PFA containers with GL threads or buttress threads.
Specifications
  • Thread:
    S 28