Order Entry
Germany
Orders LinkContactUsLinkComponent
EDTA solutions

EDTA solutions

Alkaline EDTA solution, 100 ml , for determination of exchangeable potassium in soils, Marked dropping bottle, 100 ml
Artikelnummer HACH2268732
Hersteller:   Hach
EDTA solutions
Artikelnummer HACH2268732
Hersteller:  Hach
CAS-Nummer:  
EDTA solutions
EDTA solutions

Alkaline EDTA solution, 100 ml , for determination of exchangeable potassium in soils, Marked dropping bottle, 100 ml

CAS-Nummer:  
Preis
28,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Marked dropping bottle
  • Pk:
    100 ml
  • Description:
    Alkaline EDTA solution, 100 ml , for determination of exchangeable potassium in soils

Specifications