Drum faucets
Internal flame arrester, adjustable, rigid, 3/4"
JUSU8902
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
- Safe and easy dispensing
- FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
- Thread:3/4"
- Description:Internal flame arrester, adjustable, rigid
- Seq:03