Sample cups for analysers
Sample cups for auto sampler, PP, no known solvent at room temperature, 1,2 ml, Translucent
Catalog # PERCB0510397
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
- Capacity:1,2 ml
- Colour:Translucent
- Description:Sample cups for auto sampler, PP, no known solvent at room temperature