Cleanroom boot covers, Kimtech™ A5
Wrap-around vinyl foot, White, U*, +
: Ansell
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
- Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
- Tunnelled elastic and bound seams
- Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
: PPE Cat I (EU) 2016/425
: Vacuum packed in pairs for added sterility assurance.
- Shoe size:U*
- Sterile:+
- Colour:White
- Description:Wrap-around vinyl foot
- Seq:04
- Cat. No.:113-1227
- Cleanroom class:100/ISO 5
- Garment application:Cleanroom
- Environmentally Preferable:Waste Reducing
Complementary Products
- DescriptionSterileClothing SizeColourLength