RIBOFLAVIN-DIOXOPYRIMIDINE-13C4,15N2, 99 ATOM % 13C, 98 ATOM % 15N, 97% CP 1 * 1 mg
Catalog # SIAL705292-1MG
Supplier: Merck
RIBOFLAVIN-DIOXOPYRIMIDINE-13C4,15N2, 99 ATOM % 13C, 98 ATOM % 15N, 97% CP 1 * 1 mg
Catalog # SIAL705292-1MG
Supplier: Merck
Frequently Bought Together
SHELLSOL D 40 1 * 3 L
Supplier: MARKET SOURCE PART PROCESS
Water analytical reagent
Supplier: Bernd Kraft
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
BARIUM CHLORIDE SOLUTION 0.05 MOL/L - 0. 1 * 5 L
Supplier: Bernd Kraft
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND