Order Entry
Germany
Orders LinkContactUsLinkComponent
NMR REF STD 40% 1,4-DIOXANE/BENZENE-D6, 1 * 1 items

NMR REF STD 40% 1,4-DIOXANE/BENZENE-D6, 1 * 1 items

Catalog # SIAL684228-1EA
Supplier:   Merck
NMR REF STD 40% 1,4-DIOXANE/BENZENE-D6, 1 * 1 items
Catalog # SIAL684228-1EA
Supplier:  Merck
CAS Number:  
NMR REF STD 40% 1,4-DIOXANE/BENZENE-D6, 1 * 1 items
CAS Number:  
Price
425,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.