Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®

Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 7 (3 mol/l), Glass bottle, 500 mL
Catalog # SIALS2404-500ML
Supplier:   Merck
Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®
Catalog # SIALS2404-500ML
Supplier:  Merck
CAS Number:  
Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 7 (3 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
163,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Sodium acetate buffer solution pH 7 (3 mol/l)
  • Cat. No.:
    SIALS2404-500ML
  • M-Clarity quality level:
    MQ200

Specifications