Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®
Sodium acetate buffer solution pH 7 (3 mol/l), Glass bottle, 100 mL
Artikelnummer SIALS2404-100ML
Hersteller: Merck
Buffer, sodium acetate buffer solution pH 7 (3 mol/L), Sigma-Aldrich®
Artikelnummer SIALS2404-100ML
Hersteller: Merck
CAS-Nummer:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:Sodium acetate buffer solution pH 7 (3 mol/l)
- Cat. No.:SIALS2404-100ML
- M-Clarity quality level:MQ200



