Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®
Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 500 mL
Catalog # SIALS7899-500ML
Supplier: Merck
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®
Catalog # SIALS7899-500ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:Sodium acetate buffer solution pH 5,2 (3 mol/l)
- Cat. No.:SIALS7899-500ML
- M-Clarity quality level:MQ200
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



