Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 1 L
Catalog # SIALS7899-1L
Supplier:   Merck
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®
Catalog # SIALS7899-1L
Supplier:  Merck
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 1 L

CAS Number:  
Price
143,00 €
Log in for availability
1 * 1 L
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)
  • Cat. No.:
    SIALS7899-1L
  • M-Clarity quality level:
    MQ200

Specifications