Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Methylmorpholine, Sigma-Aldrich®

4-Methylmorpholine, Sigma-Aldrich®

Glass bottle, 100 mL
Catalog # SIAL407704-100ML
Flammable
Corrosive
Irritant
Supplier:   Merck
4-Methylmorpholine, Sigma-Aldrich®
Catalog # SIAL407704-100ML
Supplier:  Merck
CAS Number:  109-02-4
4-Methylmorpholine, Sigma-Aldrich®

Glass bottle, 100 mL

CAS Number:  109-02-4
Price
83,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–66 °C
  • Density:0.922 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • UN:2535
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.