Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid, Sigma-Aldrich®

L(+)-Ascorbic acid, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALA7506-25G
Supplier:   Merck
L(+)-Ascorbic acid, Sigma-Aldrich®
Catalog # SIALA7506-25G
Supplier:  Merck
CAS Number:  50-81-7
L(+)-Ascorbic acid, Sigma-Aldrich®
L(+)-Ascorbic acid, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  50-81-7
Price
21,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications