Selenium, Sigma-Aldrich®
Glass bottle for solids, 100 g
Catalog # SIAL204307-100G
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:7782-49-2
- Formula:Se
- MDL Number:MFCD00134090
- Molecular Weight:78.96 g/mol
- Merck Index:12,08572
- Boiling Pt:∼685 °C (1013 hPa)
- Melting Pt:217 °C
- Density:4.28 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-957-4
- ADR:6.1,III
- :M-Clarity™ quality level = MQ100
- UN:3283
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



