Order Entry
Germany
Orders LinkContactUsLinkComponent
pH buffer solutions, referenced at 20 °C, Fluka™

pH buffer solutions, referenced at 20 °C, Fluka™

11,00, ±0.05, Borate, 20 °C, Plastic bottle, 1×1 L, 1 L
Catalog # 33650-1L
Supplier:   Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33650-1L
CAS Number:  
pH buffer solutions, referenced at 20 °C, Fluka™

11,00, ±0.05, Borate, 20 °C, Plastic bottle, 1×1 L, 1 L

CAS Number:  
Price
73,30 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0.05
  • Pk Info:
    1×1 L
  • pH value:
    11,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.