Order Entry
Germany
Orders LinkContactUsLinkComponent
Acetate buffer solution, Fluka™

Acetate buffer solution, Fluka™

Acetate buffer solution, 4,65, Plastic bottle, 6 L
Catalog # 31103-6X1L
Supplier:   Honeywell Chemicals
Acetate buffer solution, Fluka™
Catalog # 31103-6X1L
CAS Number:  
Acetate buffer solution, Fluka™

Acetate buffer solution, 4,65, Plastic bottle, 6 L

CAS Number:  
Price
318,00 €
Log in for availability
1 * 6 L
Product Details & Documents

This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Description:
    Acetate buffer solution
  • pH value:
    4,65

Specifications