Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Acetate buffer solution, Fluka™

Acetate buffer solution, 4,6, Plastic bottle, 500 mL
Catalog # 31048-500ML
Supplier:   Honeywell Chemicals
Acetate buffer solution, Fluka™
Catalog # 31048-500ML
CAS Number:  
Acetate buffer solution, Fluka™

Acetate buffer solution, 4,6, Plastic bottle, 500 mL

CAS Number:  
Price
48,60 €
Log in for availability
1 * 500 mL
Product Details & Documents

This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Acetate buffer solution
  • pH value:
    4,6

Specifications