Order Entry
Germany
Orders LinkContactUsLinkComponent
Acetate buffer solution, Fluka™

Acetate buffer solution, Fluka™

Acetate buffer solution, 4,6, Plastic bottle, 1 L
Catalog # 31048-1L
Supplier:   Honeywell Chemicals
Acetate buffer solution, Fluka™
Catalog # 31048-1L
CAS Number:  
Acetate buffer solution, Fluka™

Acetate buffer solution, 4,6, Plastic bottle, 1 L

CAS Number:  
Price
61,10 €
Log in for availability
1 * 1 L
Product Details & Documents

This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Acetate buffer solution
  • pH value:
    4,6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.