Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
LABORGLASKORB ROSTFREI 400X300X150MM 1 * 1 items
WCDF524-01-13
:
WDF - Funke GmbH & Co. KG
LABORGLASKORB ROSTFREI 400X300X150MM 1 * 1 items
WCDF524-01-13
:
WDF - Funke GmbH & Co. KG