Orders LinkContactUsLinkComponent
Disposable earplugs, E-A-R™ Classic™
Disposable earplugs, E-A-R™ Classic™
Earplugs, without cord, small bag of 5 pairs, Yellow, U*
Catalog # 111-0837
Supplier:   3M
Disposable earplugs, E-A-R™ Classic™
Catalog # 111-0837
Supplier:  3M
Disposable earplugs, E-A-R™ Classic™
Disposable earplugs, E-A-R™ Classic™
Earplugs, without cord, small bag of 5 pairs, Yellow, U*
Price
95,50 €
Log in for availability
1 * 250 PAAR
Product Details & Documents
These roll-down earplugs are made from a soft, energy-absorbing polymer foam which provides excellent hearing protection and all-day comfort.
  • Firm material with slow recovery time helps to achieve a good fit
  • Sweat and humidity resistant which improves comfort
  • Very economical as the earplugs can be cleaned up to three times
Suitable for use where noise is a problem either at work or leisure activities.
SNR: 28 dB
Certifications: EN 352-2.
Caution: * One size fits all.
Specifications
  • Colour:
    Yellow
  • Description:
    Earplugs, without cord, small bag of 5 pairs
  • Seq:
    03
  • Size:
    U*