Disposable earplugs, E-A-R™ Classic™
Earplugs, without cord, pillow pack, Yellow, S
Catalog # 111-1060
Supplier: 3M
Wechseln zu:
Product Details & Documents
These roll-down earplugs are made from a soft, energy-absorbing polymer foam which provides excellent hearing protection and all-day comfort.
- Firm material with slow recovery time helps to achieve a good fit
- Sweat and humidity resistant which improves comfort
- Very economical as the earplugs can be cleaned up to three times
Suitable for use where noise is a problem either at work or leisure activities.
SNR: 28 dB
Zertifizierung: EN 352-2.
Achtung! Bitte beachten: * One size fits all.
Spezifikationen:
- Colour:Yellow
- Description:Earplugs, without cord, pillow pack
- Seq:02
- Size:S