Orders LinkContactUsLinkComponent
Support clamps
Support clamps
DN 200, Schott laboratory flat flanges (PF)
Catalog # BOCH17214
Supplier:   BOCHEM
Support clamps
Catalog # BOCH17214
Supplier:  BOCHEM
Support clamps
Support clamps
DN 200, Schott laboratory flat flanges (PF)
Price
1 090,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
  • For reactors and other vessels
  • User friendly thanks to joint screws
  • With two supporting devices for 26,9 mm tubes
Specifications
  • For:
    Schott laboratory flat flanges (PF)
  • Description:
    DN 200