ContactUsLinkComponent
KIT CERAMIC 1,4MM 0,5ml tubes 50 prep, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 1,4mm ceram ic (zirconium oxide)beads in 0,5mL standard tubes is designed for soft tissue homogenizing like brain, liver, kidney an 1 * 1 KIT
Catalog # 432-3760
Supplier:  Bertin Instruments
KIT CERAMIC 1,4MM 0,5ml tubes 50 prep, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 1,4mm ceram ic (zirconium oxide)beads in 0,5mL standard tubes is designed for soft tissue homogenizing like brain, liver, kidney an 1 * 1 KIT
Catalog # 432-3760
Supplier:  Bertin Instruments
Frequently Bought Together