pH buffer solutions, referenced at 20 °C, Fluka™
10,00, ±0.05, Borate, 20 °C, Plastic bottle, 6×1 L, 6 L
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33649-6X1L
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:6 L
- Reference temperature:20 °C
- pH accuracy:±0.05
- Pk Info:6×1 L
- pH value:10,00



