Order Entry
Germany
Orders LinkContactUsLinkComponent
(S)-(+)-2-Hydroxyisovaleric acid

(S)-(+)-2-Hydroxyisovaleric acid

Glass bottle for solids, 1 g
Catalog # CAYM10007090-1
Supplier:   Cayman Chemical
(S)-(+)-2-Hydroxyisovaleric acid
Catalog # CAYM10007090-1
Supplier:  Cayman Chemical
CAS Number:  17407-55-5
(S)-(+)-2-Hydroxyisovaleric acid

Glass bottle for solids, 1 g

CAS Number:  17407-55-5
Price
78,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
  • CAS Number:17407-55-5
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066443
  • Molecular Weight:118.13 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications