Order Entry
Germany
Orders LinkContactUsLinkComponent
dNTP sets, illustra™

dNTP sets, illustra™

dNTP set (100 mM each A,C,G,T), 4×100 µmol, 1 SET
Catalog # 28-4065-52
Supplier:   Cytiva
dNTP sets, illustra™
Catalog # 28-4065-52
Supplier:  Cytiva
CAS Number:  
dNTP sets, illustra™
dNTP sets, illustra™

dNTP set (100 mM each A,C,G,T), 4×100 µmol, 1 SET

CAS Number:  
Price
795,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    dNTP set (100 mM each A,C,G,T), 4×100 µmol

Specifications