Order Entry
Germany
Orders LinkContactUsLinkComponent
 

dNTP solutions, illustra™

dATP, 100 mMol solution, 100 µmol
Catalog # 28-4065-02
Supplier:   Cytiva
dNTP solutions, illustra™
Catalog # 28-4065-02
Supplier:  Cytiva
CAS Number:  
dNTP solutions, illustra™
dNTP solutions, illustra™

dATP, 100 mMol solution, 100 µmol

CAS Number:  
Price
170,00 €
Log in for availability
1 * 100 µmol
Product Details & Documents

illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Specifications

Specifications

  • Pk:
    100 µmol
  • Description:
    dATP, 100 mMol solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.