Order Entry
Germany
Orders LinkContactUsLinkComponent
dNTP sets, illustra™

dNTP sets, illustra™

dNTP mix set (20 mM A,C,G,T), 10 µmol, 1 SET
Catalog # 28-4065-57
Supplier:   Cytiva
dNTP sets, illustra™
Catalog # 28-4065-57
Supplier:  Cytiva
CAS Number:  
dNTP sets, illustra™
dNTP sets, illustra™

dNTP mix set (20 mM A,C,G,T), 10 µmol, 1 SET

CAS Number:  
Price
147,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    dNTP mix set (20 mM A,C,G,T), 10 µmol

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.