Order Entry
Germany
Orders LinkContactUsLinkComponent
 

PIG® medium and large mobile container kits

Refill only, Oil-Only, 250,8 L
Catalog # 121-0127
Supplier:   NEW PIG
PIG® medium and large mobile container kits
Catalog # 121-0127
Supplier:  NEW PIG
PIG® medium and large mobile container kits
PIG® medium and large mobile container kits

Refill only, Oil-Only, 250,8 L

Price
486,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Fast, effective response to indoor and outdoor spills.

  • Durable, wheeled 240 and 360 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface

Weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).

 : To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.

Specifications
  • Description:
    Refill only, Oil-Only
  • Seq:
    12
  • Absorbency:
    250,8 L